FROH38A16B7B Frontier Scientific Services Inc.

2,2-Bis(tosyloxymethyl)propane-1,3-diyl bis(4-methylbenzenesulfonate)

  • MDL number MFCD00159219
  • Cas Number : 1522-89-0
  • Molecular Weight : 752.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-{[(4-methylphenyl)sulfonyl]oxy}-2,2-bis({[(4-methylphenyl)sulfonyl]oxy}methyl)propyl 4-methylbenzenesulfonate
Inchi Code 1S/C33H36O12S4/c1-25-5-13-29(14-6-25)46(34,35)42-21-33(22-43-47(36,37)30-15-7-26(2)8-16-30,23-44-48(38,39)31-17-9-27(3)10-18-31)24-45-49(40,41)32-19-11-28(4)12-20-32/h5-20H,21-24H2,1-4H3
Inchi Key KBTUOVZRXXHLOB-UHFFFAOYSA-N

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart