FROH38A16B86 Frontier Scientific Services Inc.

(3-Cyclohexylpropyl)boronic acid

  • MDL number MFCD06212395
  • Molecular Weight : 170.057

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart