FROH38A16B9A Frontier Scientific Services Inc.

5-Hydroxy-2-iodophenylboronic acid pinacol ester

  • MDL number MFCD16994433
  • Cas Number : 1256781-71-1
  • Molecular Weight : 345.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-iodo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Inchi Code 1S/C12H16BIO3/c1-11(2)12(3,4)17-13(16-11)9-7-8(15)5-6-10(9)14/h5-7,15H,1-4H3
Inchi Key QLZQKMXNNCBLSI-UHFFFAOYSA-N

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart