FROH38A16BC0 Frontier Scientific Services Inc.

Generation 2 dendron, Nitrononaester

  • MDL number MFCD13619881
  • Cas Number : 220215-00-9
  • Molecular Weight : 1469.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart