FROH38A16BCD Frontier Scientific Services Inc.

4-(4-Methylhomopiperazin-1-ylsulfonyl)phenylboronic acid pinacol ester

  • MDL number MFCD19686140
  • Molecular Weight : 380.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart