FROH38A16BD1 Frontier Scientific Services Inc.

4,4'-(2,2-Bis((4-iodophenoxy)methyl)propane-1,3-diyl)bis(oxy)bis(iodobenzene)

  • MDL number MFCD18915045
  • Cas Number : 678187-25-2
  • Molecular Weight : 944.116

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart