FROH38A16BD2 Frontier Scientific Services Inc.

3-(Morpholinosulfonyl)phenylboronic acid pinacol ester

  • MDL number MFCD19288881
  • Cas Number : 1198804-92-0
  • Molecular Weight : 353.241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart