FROH38A16BEA Frontier Scientific Services Inc.

3-(Difluoromethyl)-5-hydroxy-1-methyl-1H-pyrazole-4-carbaldeyhde

  • MDL number MFCD12827695
  • Cas Number : 1203145-73-6
  • Molecular Weight : 176.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart