FROH38A16C04 Frontier Scientific Services Inc.

3,3'-(2,2-Bis((3-oxo-3-(N-Boc-piperazin-1-yl)propoxy)methyl)propane-1,3-diyl)bis(oxy)bis(1-(N-Boc-piperazin-1-yl)propan-1-one)

  • MDL number MFCD20327969
  • Molecular Weight : 1097.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart