FROH38A16C0D Frontier Scientific Services Inc.

Weisocyanate

  • MDL number MFCD08669605
  • Cas Number : 197368-00-6
  • Molecular Weight : 441.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name di(tert-butyl) 4-(3-tert-butoxy-3-oxopropyl)-4-isocyanatoheptanedioate
Inchi Code 1S/C23H39NO7/c1-20(2,3)29-17(26)10-13-23(24-16-25,14-11-18(27)30-21(4,5)6)15-12-19(28)31-22(7,8)9/h10-15H2,1-9H3
Inchi Key OYIAIBCWADDXMC-UHFFFAOYSA-N

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart