FROH38A16C3A Frontier Scientific Services Inc.

3,3'-(2,2-Bis((3-oxo-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylamino)propoxy)methyl)propane-1,3-diyl)bis(oxy)bis(N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanamide)

  • MDL number MFCD20327983
  • Molecular Weight : 1228.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart