FROH38A16C43 Frontier Scientific Services Inc.

FMOC Aminotriester

  • MDL number MFCD16872073
  • Cas Number : 1217553-84-8
  • Molecular Weight : 637.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart