FROH38A16C4A Frontier Scientific Services Inc.

Potassium 2,4-dimethoxypyrimidine-5-trifluoroborate

  • MDL number MFCD09993667
  • Cas Number : 1111732-97-8
  • Molecular Weight : 246.036

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart