JWPH3249C1A9 J & W Pharmlab LLC

3-Chloro-5-hydroxy-benzoic acid benzyl ester

  • MDL number MFCD04114329
  • Cas Number : 245116-17-0
  • Molecular Weight : 262.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form White Solid
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0006

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0006
Availability details for
Quantity
  • Availability
  • Add to Cart