JWPH3249C234 J & W Pharmlab LLC

4-Methyl-isophthalic acid

  • MDL number MFCD09878876
  • Cas Number : 3347-99-7
  • Molecular Weight : 180.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methylisophthalic acid
Inchi Code 1S/C9H8O4/c1-5-2-3-6(8(10)11)4-7(5)9(12)13/h2-4H,1H3,(H,10,11)(H,12,13)
Inchi Key PNWSHHILERSSLF-UHFFFAOYSA-N

Description

Purity 96%
Physical Form White Solid
Product Link https://www.jwpharmlab.com/home/product/05R0304

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0304
Availability details for
Quantity
  • Availability
  • Add to Cart