JWPH3249C272 J & W Pharmlab LLC

(4-Amino-phenyl)-cyclopropyl-methanone hydrochloride

  • MDL number MFCD12913649
  • Molecular Weight : 197.6639

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/05R0381S
Availability details for
Quantity
  • Availability
  • Add to Cart