JWPH3249C2CC J & W Pharmlab LLC

4-Hydroxy-2-methoxy-benzonitrile

  • MDL number MFCD08692760
  • Molecular Weight : 149.1488

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/05R0526
Availability details for
Quantity
  • Availability
  • Add to Cart