JWPH3249C42E J & W Pharmlab LLC

N-(2-Formyl-phenyl)-acetamide

  • MDL number MFCD06739581
  • Molecular Weight : 163.1756

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form white solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/10R0232
Availability details for
Quantity
  • Availability
  • Add to Cart