JWPH3249C451 J & W Pharmlab LLC

(4-Methoxy-2-nitro-phenyl)-acetonitrile

  • MDL number MFCD07780792
  • Cas Number : 105003-90-5
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Yelllow Solid
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/10R0276

Safety Information

MSDS https://www.jwpharmlab.com/home/product/10R0276
Availability details for
Quantity
  • Availability
  • Add to Cart