JWPH3249C4E6 J & W Pharmlab LLC

3-Phenyl-cyclopent-2-enone

  • MDL number MFCD11934406
  • Cas Number : 3810-26-2
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White Solid
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/10R0479

Safety Information

MSDS https://www.jwpharmlab.com/home/product/10R0479
Availability details for
Quantity
  • Availability
  • Add to Cart