JWPH3249C674 J & W Pharmlab LLC

3-Methylaminomethyl-benzoic acid methyl ester hydrochloride

  • MDL number MFCD12913665
  • Cas Number : 1187930-01-3
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Room Temp
Physical Form White Solid
Shipping Temperature Room Temp
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0137S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0137S
Availability details for
Quantity
  • Availability
  • Add to Cart