JWPH3249C6E4 J & W Pharmlab LLC

2-Amino-3,6-difluoro-benzoic acid

  • MDL number MFCD01569542
  • Cas Number : 825654-54-4
  • Molecular Weight : 173.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form Yellow Solid
Shipping Temperature Room Temp
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0273

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0273
Availability details for
Quantity
  • Availability
  • Add to Cart