JWPH3249C784 J & W Pharmlab LLC

3-Dimethylamino-1-phenyl-propan-1-ol

  • MDL number MFCD00091135
  • Cas Number : 5554-64-3
  • Molecular Weight : 179.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form Light Brown Solid
Shipping Temperature Room Temp
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0002

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0002
Availability details for
Quantity
  • Availability
  • Add to Cart