JWPH3249C7A3 J & W Pharmlab LLC

2-Cyclohexyl-ethylamine

  • MDL number MFCD00058668
  • Cas Number : 4442-85-7
  • Molecular Weight : 127.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form Colorless Liquid
Shipping Temperature Room Temp
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0038

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0038
Availability details for
Quantity
  • Availability
  • Add to Cart