JWPH3249C7FA J & W Pharmlab LLC

3-(2-Amino-ethyl)-phenylamine dihydrochloride

  • MDL number MFCD09991570
  • Cas Number : 879666-37-2
  • Molecular Weight : 209.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form white solid
Shipping Temperature Room Temp
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0108S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0108S
Availability details for
Quantity
  • Availability
  • Add to Cart