JWPH3249C927 J & W Pharmlab LLC

(2-Fluoro-4-methyl-phenyl)-hydrazine hydrochloride

  • MDL number MFCD06738785
  • Molecular Weight : 176.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-fluoro-4-methylphenyl)hydrazine hydrochloride
Inchi Code 1S/C7H9FN2.ClH/c1-5-2-3-7(10-9)6(8)4-5;/h2-4,10H,9H2,1H3;1H
Inchi Key DFQZJMAPHJZMRH-UHFFFAOYSA-N

Description

Purity 96%
Physical Form Brown Solid
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R0348S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0348S
Availability details for
Quantity
  • Availability
  • Add to Cart