JWPH3249C997 J & W Pharmlab LLC

(R)-2,2,2-Trifluoro-1-(4-fluoro-phenyl)-ethylamine hydrochloride

  • MDL number MFCD12910559
  • Cas Number : 1391504-83-8
  • Molecular Weight : 229.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R)-2,2,2-trifluoro-1-(4-fluorophenyl)ethanamine hydrochloride
Inchi Code 1S/C8H7F4N.ClH/c9-6-3-1-5(2-4-6)7(13)8(10,11)12;/h1-4,7H,13H2;1H/t7-;/m1./s1
Inchi Key BMJQOPDJQFLGFV-OGFXRTJISA-N

Description

Purity 96%
Physical Form White Solid
Product Link https://www.jwpharmlab.com/home/product/20R0472S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R0472S
Availability details for
Quantity
  • Availability
  • Add to Cart