JWPH3249CACA J & W Pharmlab LLC

(R)-6-Fluoro-1,2,3,4-tetrahydro-naphthalen-1-ylamine hydrochloride

  • MDL number MFCD22056325
  • Molecular Weight : 201.6708

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off white solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/20R0832S
Availability details for
Quantity
  • Availability
  • Add to Cart