JWPH3249CBB8 J & W Pharmlab LLC

2,5-Dimethyl-cyclopentylamine hydrochloride

  • MDL number MFCD21602652
  • Molecular Weight : 149.6631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form White Solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/20R1107S
Availability details for
Quantity
  • Availability
  • Add to Cart