JWPH3249CBE4 J & W Pharmlab LLC

Chroman-8-ylamine hydrochloride

  • MDL number MFCD26407866
  • Molecular Weight : 185.6529

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/20R1160S
Availability details for
Quantity
  • Availability
  • Add to Cart