JWPH3249CBEC J & W Pharmlab LLC

(S)-(2-Amino-4-methyl-pentyl)-carbamic acid tert-butyl ester

  • MDL number MFCD22418541
  • Cas Number : 1217317-66-2
  • Molecular Weight : 216.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form Light Yellow Liquid
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1167

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1167
Availability details for
Quantity
  • Availability
  • Add to Cart