JWPH3249D0A6 J & W Pharmlab LLC

5-Pyridin-3-yl-1H-indazole

  • MDL number MFCD04114676
  • Molecular Weight : 195.2232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form yellow solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/45R0022
Availability details for
Quantity
  • Availability
  • Add to Cart