JWPH3249D28F J & W Pharmlab LLC

6-fluoro-3-(1,2,3,6-tetrahydro-pyridin-4-yl)-1H-indole

  • MDL number MFCD04114772
  • Molecular Weight : 216.2575

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form orange solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/48R0085
Availability details for
Quantity
  • Availability
  • Add to Cart