JWPH3249D5EC J & W Pharmlab LLC

1-Chloro-isoquinoline-4-carboxylic acid methyl ester

  • MDL number MFCD00140353
  • Molecular Weight : 221.6427

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Physical Form White Solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/50R0280
Availability details for
Quantity
  • Availability
  • Add to Cart