JWPH3249D611 J & W Pharmlab LLC

6-Chloro-3,4-dihydro-2H-[2,7]naphthyridin-1-one

  • MDL number MFCD18250874
  • Cas Number : 1260664-25-2
  • Molecular Weight : 182.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Room Temp
Physical Form off white solid
Shipping Temperature Room Temp
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/50R0321

Safety Information

MSDS https://www.jwpharmlab.com/home/product/50R0321
Availability details for
Quantity
  • Availability
  • Add to Cart