JWPH3249DEA2 J & W Pharmlab LLC

1-(3-Bromo-phenyl)-1H-pyrazole

  • MDL number MFCD06797472
  • Cas Number : 294877-33-1
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Room Temp
Physical Form brown solid
Shipping Temperature Room Temp
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0376

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0376
Availability details for
Quantity
  • Availability
  • Add to Cart