JWPH3249E836 J & W Pharmlab LLC

2,7-Diaza-spiro[4.4]nonane dihydrochloride

  • MDL number MFCD10700113
  • Cas Number : 1394122-72-5
  • Molecular Weight : 199.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form White Solid
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0299S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0299S
Availability details for
Quantity
  • Availability
  • Add to Cart