JWPH3249ECCB J & W Pharmlab LLC

4-Ethyl-thiazole-2-carboxylic acid ethyl ester

  • MDL number MFCD11520453
  • Molecular Weight : 185.2404

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Physical Form yellow liquid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/90R0380
Availability details for
Quantity
  • Availability
  • Add to Cart