JWPH3249ED95 J & W Pharmlab LLC

4-Hydrazino-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD16496498
  • Molecular Weight : 215.2948

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/60R1406
Availability details for
Quantity
  • Availability
  • Add to Cart