JWPH37C05091 J & W Pharmlab LLC

2-Morpholin-4-yl-propionic acid

  • MDL number MFCD03821872
  • Cas Number : 25245-81-2
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/53R0192

Safety Information

MSDS https://www.jwpharmlab.com/home/product/53R0192
Availability details for
Quantity
  • Availability
  • Add to Cart