JWPH37C05097 J & W Pharmlab LLC

(4R)-N-Cbz-4-hydroxy-azepane

  • MDL number MFCD11975381
  • Molecular Weight : 249.309

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/60R0732
Availability details for
Quantity
  • Availability
  • Add to Cart