JWPH37C05099 J & W Pharmlab LLC

(3R,4R)-(1-Benzyl-3-hydroxy-piperidin-4-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD18914359
  • Molecular Weight : 306.404

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/60R0782
Availability details for
Quantity
  • Availability
  • Add to Cart