JWPH37C0509C J & W Pharmlab LLC

2,3-Difluoro-5-methoxy-benzoic acid

  • MDL number MFCD28402910
  • Molecular Weight : 188.1304

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/05R0770
Availability details for
Quantity
  • Availability
  • Add to Cart