JWPH37C0509F J & W Pharmlab LLC

(4-Cyano-2-nitro-phenoxy)-acetic acid ethyl ester

  • MDL number MFCD16439454
  • Molecular Weight : 250.2104

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/05R0910
Availability details for
Quantity
  • Availability
  • Add to Cart