JWPH37C050A1 J & W Pharmlab LLC

Pyridine-4-sulfonic acid methylamide

  • MDL number MFCD26599494
  • Molecular Weight : 172.2014

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/69R1124
Availability details for
Quantity
  • Availability
  • Add to Cart