JWPH37C050A2 J & W Pharmlab LLC

Pyridine-4-sulfonic acid cyclopropylamide

  • MDL number MFCD28098150
  • Molecular Weight : 198.2392

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/69R1125
Availability details for
Quantity
  • Availability
  • Add to Cart