JWPH37C050A3 J & W Pharmlab LLC

2-Methyl-5,7-dinitro-quinolin-8-ol

  • MDL number MFCD13191630
  • Cas Number : 38543-75-8
  • Molecular Weight : 249.1824

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Yellow Solid
Shipping Temperature RT
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/69R1366
Availability details for
Quantity
  • Availability
  • Add to Cart