JWPH37C050A6 J & W Pharmlab LLC

(5-Methoxy-pyridin-3-yl)-methyl-amine

  • MDL number MFCD26903113
  • Molecular Weight : 138.1688

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/69R1162
Availability details for
Quantity
  • Availability
  • Add to Cart