JWPH37C050A7 J & W Pharmlab LLC

(2,6-Dichloro-pyrimidin-4-yl)-acetic acid methyl ester

  • MDL number MFCD13705338
  • Molecular Weight : 221.0426

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/70R0388
Availability details for
Quantity
  • Availability
  • Add to Cart