JWPH37C050AB J & W Pharmlab LLC

2,2-Dibromo-1-methyl-cyclopropanecarboxylic acid

  • MDL number MFCD00760330
  • Cas Number : 5365-21-9
  • Molecular Weight : 257.9092

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form Off White Solid
Shipping Temperature RT
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/05R0912
Availability details for
Quantity
  • Availability
  • Add to Cart